Product Details

SNP ID
rs149642401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149376858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCACCATCTCCTCGATGTTCCT[C/T]TCATACTCCTCCATGCGGGCATACG
Phenotype
MIM: 604627
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17B PubMed Links

Gene Details

Gene
IL17B
Gene Name
interleukin 17B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317987.1 562 Silent Mutation GAA,GAG E11E NP_001304916.1
NM_014443.2 562 Silent Mutation GAA,GAG E63E NP_055258.1
XM_011537622.2 562 Silent Mutation GAA,GAG E11E XP_011535924.1
XM_017009346.1 562 Silent Mutation GAA,GAG E128E XP_016864835.1
XM_017009347.1 562 Silent Mutation GAA,GAG E11E XP_016864836.1
XM_017009348.1 562 Silent Mutation GAA,GAG E11E XP_016864837.1
Gene
PCYOX1L
Gene Name
prenylcysteine oxidase 1 like
There are no transcripts associated with this gene.

View Full Product Details