Product Details

SNP ID
rs149642459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177457698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGGAGGCGGAGCCTTGGCACA[C/G]AGCTCTTCCGATTGGGCAAACTCCT
Phenotype
MIM: 126660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBN1 PubMed Links

Gene Details

Gene
DBN1
Gene Name
drebrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004395.3 2124 Silent Mutation CTC,CTG L612L NP_004386.2
NM_080881.2 2124 Silent Mutation CTC,CTG L614L NP_543157.1
XM_005265827.3 2124 Silent Mutation CTC,CTG L658L XP_005265884.1
XM_006714826.2 2124 Silent Mutation CTC,CTG L595L XP_006714889.1
XM_011534446.2 2124 Silent Mutation CTC,CTG L660L XP_011532748.1
XM_011534447.2 2124 Silent Mutation CTC,CTG L647L XP_011532749.1
XM_017009139.1 2124 Silent Mutation CTC,CTG L660L XP_016864628.1
XM_017009140.1 2124 Silent Mutation CTC,CTG L614L XP_016864629.1
Gene
PRR7
Gene Name
proline rich 7 (synaptic)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174101.1 2124 Intron NP_001167572.1
NM_001174102.1 2124 Intron NP_001167573.1
NM_030567.4 2124 Intron NP_085044.2
XM_011534662.1 2124 Intron XP_011532964.1
XM_011534663.2 2124 Intron XP_011532965.1
XM_017009896.1 2124 Intron XP_016865385.1
XM_017009897.1 2124 Intron XP_016865386.1
Gene
PRR7-AS1
Gene Name
PRR7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details