Product Details

SNP ID
rs149655356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81987632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACGTTATTGTGCAGAATTCATTA[A/G]ACATTCACAACAGATAGGTGATAGT
Phenotype
MIM: 610800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG10 PubMed Links

Gene Details

Gene
ATG10
Gene Name
autophagy related 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131028.1 360 Missense Mutation AAA,AGA K21R NP_001124500.1
NM_031482.4 360 Missense Mutation AAA,AGA K21R NP_113670.1
XM_005248610.4 360 Missense Mutation AAA,AGA K21R XP_005248667.1
XM_005248611.4 360 Intron XP_005248668.1
XM_005248612.3 360 Missense Mutation AAA,AGA K21R XP_005248669.1
XM_011543660.2 360 Intron XP_011541962.1
XM_011543661.2 360 Intron XP_011541963.1
XM_017009944.1 360 Missense Mutation AAA,AGA K21R XP_016865433.1

View Full Product Details