Product Details

SNP ID
rs149910694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178205865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAATGTTCGTTGGTGGCCTGA[C/G]CTGGGATACTAGCAAAAAAGATTTA
Phenotype
MIM: 602688 MIM: 614683
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPAB PubMed Links

Gene Details

Gene
HNRNPAB
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004499.3 490 Missense Mutation ACC,AGC T78S NP_004490.2
NM_031266.2 490 Missense Mutation ACC,AGC T78S NP_112556.2
Gene
PHYKPL
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
There are no transcripts associated with this gene.

View Full Product Details