Product Details

SNP ID
rs149938723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:66596971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCTTCCCGGAGGAGCTGTGCCG[C/T]CCGCGCCCCGGGGCAGCAGCGCGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST4 PubMed Links

Gene Details

Gene
MAST4
Gene Name
microtubule associated serine/threonine kinase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164664.1 624 Missense Mutation CCC,TCC P106S NP_001158136.1
NM_001290226.1 624 Intron NP_001277155.1
NM_001290227.1 624 Intron NP_001277156.1
NM_001290228.1 624 Missense Mutation CCC,TCC P106S NP_001277157.1
NM_001297651.1 624 Intron NP_001284580.1
NM_015183.2 624 Intron NP_055998.1
NM_198828.2 624 Missense Mutation CCC,TCC P106S NP_942123.1
XM_006714606.3 624 Intron XP_006714669.1
XM_006714610.2 624 Intron XP_006714673.1
XM_011543382.2 624 Missense Mutation CCC,TCC P106S XP_011541684.1
XM_011543384.2 624 Intron XP_011541686.1
XM_011543385.2 624 Intron XP_011541687.1
XM_011543386.2 624 Intron XP_011541688.1
XM_017009447.1 624 Missense Mutation CCC,TCC P106S XP_016864936.1
XM_017009448.1 624 Missense Mutation CCC,TCC P106S XP_016864937.1
XM_017009449.1 624 Intron XP_016864938.1
XM_017009450.1 624 Intron XP_016864939.1
XM_017009451.1 624 Intron XP_016864940.1
XM_017009452.1 624 Intron XP_016864941.1
XM_017009453.1 624 Intron XP_016864942.1

View Full Product Details