Product Details

SNP ID
rs149939829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:103548877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGATTTTAAAAATTTAATGTA[G/T]TATTACATTCATTTCCAGTAGAATA
Phenotype
MIM: 609232
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC102467212 PubMed Links

Gene Details

Gene
LOC102467212
Gene Name
uncharacterized LOC102467212
There are no transcripts associated with this gene.

Gene
NUDT12
Gene Name
nudix hydrolase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300741.1 3424 UTR 3 NP_001287670.1
NM_031438.3 3424 UTR 3 NP_113626.1
XM_005272095.1 3424 UTR 3 XP_005272152.1
XM_005272097.3 3424 UTR 3 XP_005272154.1

View Full Product Details