Product Details

SNP ID
rs150152346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148314109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTATATGACACAGGTGGACTGC[A/G]GCATTTACAAGAAGTATCCAGTGGT
Phenotype
MIM: 613511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102546294 PubMed Links

Gene Details

Gene
LOC102546294
Gene Name
uncharacterized LOC102546294
There are no transcripts associated with this gene.

Gene
SPINK7
Gene Name
serine peptidase inhibitor, Kazal type 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032566.2 154 Missense Mutation AGC,GGC S33G NP_115955.1
Gene
SPINK9
Gene Name
serine peptidase inhibitor, Kazal type 9
There are no transcripts associated with this gene.

View Full Product Details