Product Details

SNP ID
rs150223677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157087198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGGGCTCCTCCACTTCATATAC[A/G]TTCTCTTCAATGGTATAGATGTTTT
Phenotype
MIM: 606652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAVCR2 PubMed Links

Gene Details

Gene
HAVCR2
Gene Name
hepatitis A virus cellular receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032782.4 1064 Silent Mutation AAC,AAT N270N NP_116171.3

View Full Product Details