Product Details
- SNP ID
-
rs150275549
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:39285227 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTCATTGGGGAACTCTAGGGCT[A/G]GCAATCCTAGAGAAAACAAATAAGT
- Phenotype
-
MIM: 120940
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C9
PubMed Links
Gene Details
- Gene
- C9
- Gene Name
- complement component 9
There are no transcripts associated with this gene.
View Full Product Details