Product Details

SNP ID
rs150350809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132193012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGAAGGACAGAAAAGGATGTC[A/T]GTCAACTTCTGTTGATCCACAAGGT
Phenotype
MIM: 600608
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
P4HA2 PubMed Links

Gene Details

Gene
P4HA2
Gene Name
prolyl 4-hydroxylase subunit alpha 2
There are no transcripts associated with this gene.

Gene
P4HA2-AS1
Gene Name
P4HA2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details