Product Details

SNP ID
rs150441571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55107909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCCATGCTATGACCCAGCCACA[C/T]GCGAAGGTGACCTTAAACTTTTACA
Phenotype
MIM: 140050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
There are no transcripts associated with this gene.

Gene
GZMA
Gene Name
granzyme A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006144.3 368 Missense Mutation CGC,TGC R111C NP_006135.1

View Full Product Details