Product Details

SNP ID
rs150581567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131162590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTACACTTTCATCATCATCTTC[C/T]GCCACAGAAATCTGGGATATATGTT
Phenotype
MIM: 601314 MIM: 615831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HINT1 PubMed Links

Gene Details

Gene
HINT1
Gene Name
histidine triad nucleotide binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005340.6 334 Intron NP_005331.1
XM_017009410.1 334 Silent Mutation GCA,GCG A66A XP_016864899.1
Gene
LYRM7
Gene Name
LYR motif containing 7
There are no transcripts associated with this gene.

View Full Product Details