Product Details

SNP ID
rs150867395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149860296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCTTGATATTGGAATCTGGGT[A/G]ATGGGTATGTGGGGATTCAACGTAC
Phenotype
MIM: 180071 MIM: 608886
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDE6A PubMed Links

Gene Details

Gene
PDE6A
Gene Name
phosphodiesterase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000440.2 2371 UTR 3 NP_000431.2
XM_011537650.2 2371 UTR 3 XP_011535952.1
XM_011537651.2 2371 UTR 3 XP_011535953.1
XM_011537652.1 2371 UTR 3 XP_011535954.1
XM_011537653.2 2371 UTR 3 XP_011535955.1
XM_011537654.2 2371 UTR 3 XP_011535956.1
XM_017009572.1 2371 UTR 3 XP_016865061.1
Gene
PPARGC1B
Gene Name
PPARG coactivator 1 beta
There are no transcripts associated with this gene.

View Full Product Details