Product Details

SNP ID
rs150882676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150710265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCACTCAGGTTTTCTTTGGGTT[A/C]TTTTCCAGGTCCCAAACAGAACAAC
Phenotype
MIM: 614758 MIM: 612430
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCTN4 PubMed Links

Gene Details

Gene
DCTN4
Gene Name
dynactin subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135643.1 2406 UTR 3 NP_001129115.1
NM_001135644.1 2406 UTR 3 NP_001129116.1
NM_016221.3 2406 UTR 3 NP_057305.1
XM_005268457.1 2406 UTR 3 XP_005268514.1
XM_011537644.1 2406 UTR 3 XP_011535946.1
XM_011537645.1 2406 UTR 3 XP_011535947.1
XM_011537646.1 2406 UTR 3 XP_011535948.1
XM_011537647.1 2406 UTR 3 XP_011535949.1
Gene
RBM22
Gene Name
RNA binding motif protein 22
There are no transcripts associated with this gene.

View Full Product Details