Product Details

SNP ID
rs150936707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:45262204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGTCTGGAAATCAGAGTGGAC[A/C]CCTCATGGGGCAGCGAGGGCTGCGA
Phenotype
MIM: 602780
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HCN1 PubMed Links

Gene Details

Gene
HCN1
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021072.3 2415 Missense Mutation GGG,GTG G797V NP_066550.2

View Full Product Details