Product Details

SNP ID
rs151173760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1320679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAAGCAGGCCAGCCGGTGAGAC[A/G]TGGGCATGGTGATGATGAAGGCAAA
Phenotype
MIM: 612585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1L PubMed Links

Gene Details

Gene
CLPTM1L
Gene Name
CLPTM1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030782.4 1662 Missense Mutation ACG,ATG T490M NP_110409.2
XM_011514144.2 1662 Missense Mutation ACG,ATG T489M XP_011512446.1

View Full Product Details