Product Details

SNP ID
rs151175650
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140807672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGAGGGCGCCTCGGATGCAGATA[C/T]CGGGGAGAACGCCCTGCTCACTTAC
Phenotype
MIM: 606307 MIM: 606308 MIM: 606309 MIM: 606310
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs3822346] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018900.3 599 Intron NP_061723.1
NM_031410.2 599 Intron NP_113598.1
NM_031411.2 599 Intron NP_113599.1
Gene
PCDHA2
Gene Name
protocadherin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018905.2 599 Intron NP_061728.1
NM_031495.1 599 Intron NP_113683.1
NM_031496.1 599 Intron NP_113684.1
Gene
PCDHA3
Gene Name
protocadherin alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018906.2 599 Intron NP_061729.1
NM_031497.1 599 Intron NP_113685.1
Gene
PCDHA4
Gene Name
protocadherin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018907.3 599 Missense Mutation ACC,ATC T162I NP_061730.1
NM_031500.2 599 Missense Mutation ACC,ATC T162I NP_113688.1

View Full Product Details