Product Details

SNP ID
rs151181060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141100901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCAGTCATCAGACAGGTGATTT[G/T]CTCCTGAATGAGAAATTGGACCGGG
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
There are no transcripts associated with this gene.

Gene
PCDHB3
Gene Name
protocadherin beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018937.4 660 Missense Mutation TTG,TTT L84F NP_061760.1

View Full Product Details