Product Details
- SNP ID
-
hCV171762937
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141095293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGACTTGGATGTAGGAACCAACA[C/G]TCTCCAAAATTACACAATCAGTCCC
- Phenotype
-
MIM: 606328
MIM: 606329
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHB2
PubMed Links
Gene Details
- Gene
- PCDHB2
- Gene Name
- protocadherin beta 2
There are no transcripts associated with this gene.
- Gene
- PCDHB3
- Gene Name
- protocadherin beta 3
There are no transcripts associated with this gene.
View Full Product Details