Product Details

SNP ID
rs151327162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123346092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAAATGAGTTAAACTGGTTACA[A/G]TTGGTCTCAACTTTTAAGAATTTAC
Phenotype
MIM: 613446
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP120 PubMed Links

Gene Details

Gene
CEP120
Gene Name
centrosomal protein 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166226.1 3838 UTR 3 NP_001159698.1
NM_153223.3 3838 UTR 3 NP_694955.2
XM_005271901.4 3838 UTR 3 XP_005271958.1
XM_011543185.2 3838 UTR 3 XP_011541487.1
XM_011543186.2 3838 UTR 3 XP_011541488.1
XM_017009085.1 3838 UTR 3 XP_016864574.1

View Full Product Details