Product Details

SNP ID
rs137955038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:136924379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGACCAGAAAGCAAAGCAACTAC[A/G]AGGACCTGGAGGCCCAGCCTCGGGG
Phenotype
MIM: 612519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35D3 PubMed Links

Gene Details

Gene
SLC35D3
Gene Name
solute carrier family 35 member D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008783.2 1122 Missense Mutation AAG,GAG K312E NP_001008783.1

View Full Product Details