Product Details

SNP ID
rs137982429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26271004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCCCCACGAATGCGGCGAGC[C/G]AGCTGAATGTCCTTGGGCATGATAG
Phenotype
MIM: 602807 MIM: 602815
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIST1H2BI PubMed Links

Gene Details

Gene
HIST1H2BI
Gene Name
histone cluster 1, H2bi
There are no transcripts associated with this gene.

Gene
HIST1H3G
Gene Name
histone cluster 1, H3g
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003534.2 381 Silent Mutation CTC,CTG L127L NP_003525.1

View Full Product Details