Product Details

SNP ID
rs137982888
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73363213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTATTTGGGAGGCCTTCTTACCA[A/G]GAGGACACAATCAAGATGATCATGA
Phenotype
MIM: 611111 MIM: 611687 MIM: 611689
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DPPA5 PubMed Links
Additional Information
For this assay, SNP(s) [rs564533] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DPPA5
Gene Name
developmental pluripotency associated 5
There are no transcripts associated with this gene.

Gene
KHDC3L
Gene Name
KH domain containing 3 like, subcortical maternal complex member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017361.2 341 Silent Mutation CAA,CAG Q96Q NP_001017361.1
Gene
OOEP
Gene Name
oocyte expressed protein
There are no transcripts associated with this gene.

Gene
OOEP-AS1
Gene Name
OOEP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details