Product Details

SNP ID
rs138053809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:157298266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCTCAGGAGCGATTCTAAAA[C/T]GATTTTTACTCAGTCAGGCTTTATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHAL6FP PubMed Links

Gene Details

Gene
LDHAL6FP
Gene Name
lactate dehydrogenase A like 6F, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM242
Gene Name
transmembrane protein 242
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018452.5 Intron NP_060922.2

View Full Product Details