Product Details

SNP ID
rs138097862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30948868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAAGGGAAAGACCCAATGATCC[A/G]GAACCAGCGCTCTGTTGATCCTGCT
Phenotype
MIM: 613928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080870.3 404 Missense Mutation CAG,CGG Q135R NP_543146.2

View Full Product Details