Product Details

SNP ID
rs138132800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36131296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCCATCGACAAGCGGTCAGGG[A/G]AGAAGGTGGCCATCAAGAAGCTGAG
Phenotype
MIM: 602899 MIM: 600289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK13 PubMed Links

Gene Details

Gene
MAPK13
Gene Name
mitogen-activated protein kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002754.4 244 Missense Mutation AAG,GAG K49E NP_002745.1
Gene
MAPK14
Gene Name
mitogen-activated protein kinase 14
There are no transcripts associated with this gene.

View Full Product Details