Product Details

SNP ID
rs138242272
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34293497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTCCAGCACTTCAGTGACAAT[A/G]AGCACATAGACATACGTCCTGTGCT
Phenotype
MIM: 609228
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NUDT3 PubMed Links

Gene Details

Gene
NUDT3
Gene Name
nudix hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006703.3 745 Silent Mutation CTC,CTT L98L NP_006694.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 745 Silent Mutation CTC,CTT L217L NP_001189399.1

View Full Product Details