Product Details

SNP ID
rs138245764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:134171125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATTTGGTTTCAGCTGGAGAGG[C/T]TTGTTCAGAATGTTGTCGTACATTT
Phenotype
MIM: 602958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGK1 PubMed Links

Gene Details

Gene
SGK1
Gene Name
serum/glucocorticoid regulated kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143676.1 1070 Silent Mutation AAA,AAG K407K NP_001137148.1
NM_001143677.1 1070 Silent Mutation AAA,AAG K340K NP_001137149.1
NM_001143678.1 1070 Silent Mutation AAA,AAG K326K NP_001137150.1
NM_001291995.1 1070 Silent Mutation AAA,AAG K268K NP_001278924.1
NM_005627.3 1070 Silent Mutation AAA,AAG K312K NP_005618.2
XM_011536071.1 1070 Silent Mutation AAA,AAG K407K XP_011534373.1

View Full Product Details