Product Details

SNP ID
rs138353816
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125986561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGGGCAGCTGCAATGGCGCTGT[C/T]GTGTACCCTTAACAGGTATCTGCTC
Phenotype
MIM: 609998
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HINT3 PubMed Links

Gene Details

Gene
HINT3
Gene Name
histidine triad nucleotide binding protein 3
There are no transcripts associated with this gene.

Gene
TRMT11
Gene Name
tRNA methyltransferase 11 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031712.2 33 Intron NP_001026882.2
XM_017011168.1 33 UTR 5 XP_016866657.1
XM_017011169.1 33 UTR 5 XP_016866658.1

View Full Product Details