Product Details

SNP ID
rs138378309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149597693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGATATTAAGTTGGATTATACC[A/T]AATGAAGCTCAGCTATTTAAAGATC
Phenotype
MIM: 606696
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GINM1 PubMed Links

Gene Details

Gene
GINM1
Gene Name
glycoprotein integral membrane 1
There are no transcripts associated with this gene.

Gene
KATNA1
Gene Name
katanin catalytic subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204076.1 Intron NP_001191005.1
NM_007044.3 Intron NP_008975.1
XM_005266804.2 Intron XP_005266861.1
XM_005266805.3 Intron XP_005266862.1
XM_017010207.1 Intron XP_016865696.1
XM_017010208.1 Intron XP_016865697.1
XM_017010209.1 Intron XP_016865698.1
XM_017010210.1 Intron XP_016865699.1
Gene
RPS18P9
Gene Name
ribosomal protein S18 pseudogene 9
There are no transcripts associated with this gene.

View Full Product Details