Product Details

SNP ID
rs138395717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33315710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGGGGGCATAGGAAGAGGGAG[C/T]TGGCCCCCCTGAGTCATCAAGACCT
Phenotype
MIM: 603186 MIM: 601962 MIM: 611439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAXX PubMed Links

Gene Details

Gene
DAXX
Gene Name
death domain associated protein
There are no transcripts associated with this gene.

Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
ZBTB22
Gene Name
zinc finger and BTB domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145338.1 1341 Missense Mutation ACT,GCT T403A NP_001138810.1
NM_005453.4 1341 Missense Mutation ACT,GCT T403A NP_005444.4

View Full Product Details