Product Details
- SNP ID
-
rs138592460
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:96891249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGCAGTCCAGGTTTCTGGGAAA[A/C]GCTTATGATTATTGAGAAGTGTCAA
- Phenotype
-
MIM: 611776
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL32
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11402,rs34213186] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLHL32
- Gene Name
- kelch like family member 32
There are no transcripts associated with this gene.
- Gene
- NDUFAF4
- Gene Name
- NADH:ubiquinone oxidoreductase complex assembly factor 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014165.3 |
|
Intron |
|
|
NP_054884.1 |
View Full Product Details