Product Details

SNP ID
rs138600096
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32949723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAAGGCCTGGAAGCTGAGTCC[A/G]TCGACAGCTGAGACAAAAGTAGGCC
Phenotype
MIM: 142855 MIM: 142856
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DMA PubMed Links
Additional Information
For this assay, SNP(s) [rs6926628] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-DMA
Gene Name
major histocompatibility complex, class II, DM alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006120.3 626 Silent Mutation GAA,GAG E180E NP_006111.2
Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
There are no transcripts associated with this gene.

View Full Product Details