Product Details

SNP ID
rs138655717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24429068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCTATAGACGTGAAGTGCCCC[A/G]GAGAGTCGGGCTCCCAAAGAACTCC
Phenotype
MIM: 602515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPLD1 PubMed Links

Gene Details

Gene
GPLD1
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001503.3 2598 Silent Mutation TCC,TCT S829S NP_001494.2
XM_011514509.2 2598 Intron XP_011512811.1
XM_017010753.1 2598 Silent Mutation TCC,TCT S839S XP_016866242.1
XM_017010754.1 2598 Silent Mutation TCC,TCT S666S XP_016866243.1
Gene
MRS2
Gene Name
MRS2, magnesium transporter
There are no transcripts associated with this gene.

View Full Product Details