Product Details

SNP ID
rs138718587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35944239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTCCAGCTCCAAATCCAACTC[C/T]GACTCCTCTTCTACAGGCTGTTGGA
Phenotype
MIM: 608480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC26A8 PubMed Links

Gene Details

Gene
SLC26A8
Gene Name
solute carrier family 26 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193476.1 2709 Silent Mutation TCA,TCG S858S NP_001180405.1
NM_052961.3 2709 Silent Mutation TCA,TCG S858S NP_443193.1
NM_138718.2 2709 Silent Mutation TCA,TCG S753S NP_619732.2
XM_011514294.2 2709 Silent Mutation TCA,TCG S832S XP_011512596.1
XM_017010235.1 2709 Silent Mutation TCA,TCG S858S XP_016865724.1

View Full Product Details