Product Details
- SNP ID
-
rs138720439
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28995513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACAATCCCTGCATTGATAGGGCT[C/T]CTCCCCTGTATGCTCTCGTTCATGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HCG16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9295783] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCG16
- Gene Name
- HLA complex group 16 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- ZNF311
- Gene Name
- zinc finger protein 311
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010877.3 |
1952 |
Missense Mutation |
AAG,GAG |
K497E |
NP_001010877.2 |
NM_001318534.1 |
1952 |
Missense Mutation |
AAG,GAG |
K405E |
NP_001305463.1 |
NM_001318535.1 |
1952 |
Missense Mutation |
AAG,GAG |
K405E |
NP_001305464.1 |
XM_005249019.3 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_005249076.1 |
XM_006715067.2 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_006715130.1 |
XM_011514510.1 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_011512812.1 |
XM_011514511.2 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_011512813.1 |
XM_011514512.1 |
1952 |
Missense Mutation |
AAG,GAG |
K522E |
XP_011512814.1 |
XM_011514513.1 |
1952 |
Missense Mutation |
AAG,GAG |
K521E |
XP_011512815.1 |
XM_011514514.1 |
1952 |
Missense Mutation |
AAG,GAG |
K505E |
XP_011512816.1 |
XM_011514516.1 |
1952 |
Missense Mutation |
AAG,GAG |
K449E |
XP_011512818.1 |
XM_011514517.1 |
1952 |
Missense Mutation |
AAG,GAG |
K405E |
XP_011512819.1 |
XM_011514518.1 |
1952 |
Missense Mutation |
AAG,GAG |
K405E |
XP_011512820.1 |
XM_017010756.1 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_016866245.1 |
XM_017010757.1 |
1952 |
Missense Mutation |
AAG,GAG |
K530E |
XP_016866246.1 |
XM_017010758.1 |
1952 |
Missense Mutation |
AAG,GAG |
K497E |
XP_016866247.1 |
XM_017010759.1 |
1952 |
Missense Mutation |
AAG,GAG |
K497E |
XP_016866248.1 |
XM_017010760.1 |
1952 |
Missense Mutation |
AAG,GAG |
K449E |
XP_016866249.1 |
XM_017010761.1 |
1952 |
Missense Mutation |
AAG,GAG |
K405E |
XP_016866250.1 |
View Full Product Details