Product Details

SNP ID
rs138882886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41921574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTCGAGAAAAATGCCCAAATT[C/T]AAGGCGGCCCGTGGGGTGGGGGGTC
Phenotype
MIM: 603871 MIM: 612915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BYSL PubMed Links

Gene Details

Gene
BYSL
Gene Name
bystin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004053.3 348 Silent Mutation TTC,TTT F4F NP_004044.3
Gene
MED20
Gene Name
mediator complex subunit 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305455.1 348 Intron NP_001292384.1
NM_001305456.1 348 Intron NP_001292385.1
NM_001305457.1 348 Intron NP_001292386.1
NM_004275.4 348 Intron NP_004266.2

View Full Product Details