Product Details

SNP ID
rs138955257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39105569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCTTTCTTCTCTTCAGGGCCTC[A/G]TGTCCCGACGTACATCCAATAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAYSD1 PubMed Links

Gene Details

Gene
SAYSD1
Gene Name
SAYSVFN motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304793.1 512 Nonsense Mutation CGA,TGA R72* NP_001291722.1
NM_018322.2 512 Nonsense Mutation CGA,TGA R139* NP_060792.1

View Full Product Details