Product Details

SNP ID
rs138983902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:5999035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACACCAGGAGCACCGGGAACG[C/T]CGGGAGCAGGGACCCCGCCGCCCCG
Phenotype
MIM: 607409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRN1 PubMed Links

Gene Details

Gene
NRN1
Gene Name
neuritin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278710.1 1412 Missense Mutation NP_001265639.1
NM_001278711.1 1412 Missense Mutation NP_001265640.1
NM_016588.2 1412 Missense Mutation NP_057672.1
XM_006715106.3 1412 Missense Mutation XP_006715169.1
XM_006715107.3 1412 Missense Mutation XP_006715170.1

View Full Product Details