Product Details

SNP ID
rs139018301
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24667289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACCAGCATGACTCAGTCTCTGC[A/G]GGAGGTGATAAAGGCCATGACCAAG
Phenotype
MIM: 615652 MIM: 605764
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACOT13 PubMed Links

Gene Details

Gene
ACOT13
Gene Name
acyl-CoA thioesterase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160094.1 255 UTR 5 NP_001153566.1
NM_018473.3 255 Missense Mutation CAG,CGG Q9R NP_060943.1
Gene
TDP2
Gene Name
tyrosyl-DNA phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016614.2 255 Intron NP_057698.2

View Full Product Details