Product Details

SNP ID
rs139045661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:160217445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCTCTCTTCTTAGTTCAATGGA[A/T]TGTCTAGTTTCTGAACTTGGAGGTA
Phenotype
MIM: 602608
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC22A2 PubMed Links

Gene Details

Gene
SLC22A2
Gene Name
solute carrier family 22 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003058.3 1829 Missense Mutation AAT,ATT N552I NP_003049.2

View Full Product Details