Product Details
- SNP ID
-
rs139057187
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29462216 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGGCATCTGTGGCCTGGGTTAT[A/G]AGTCTGGTTCAATCGATAGTCCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OR2H1
PubMed Links
Gene Details
- Gene
- OR2H1
- Gene Name
- olfactory receptor family 2 subfamily H member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318014.1 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
NP_001304943.1 |
NM_001318022.1 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
NP_001304951.1 |
NM_030883.4 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
NP_112145.1 |
XM_011514470.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512772.1 |
XM_011514472.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512774.1 |
XM_011514473.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512775.1 |
XM_011514479.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512781.1 |
XM_011514483.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512785.1 |
XM_011514484.2 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_011512786.1 |
XM_017010737.1 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_016866226.1 |
XM_017010738.1 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_016866227.1 |
XM_017010739.1 |
2189 |
Missense Mutation |
ATA,ATG |
I149M |
XP_016866228.1 |
XM_017010740.1 |
2189 |
Missense Mutation |
ATA,ATG |
I71M |
XP_016866229.1 |
- Gene
- OR5V1
- Gene Name
- olfactory receptor family 5 subfamily V member 1
There are no transcripts associated with this gene.
View Full Product Details