Product Details

SNP ID
rs139062302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111263918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCACTGTCATGATGTCTATCTT[C/T]GGTGTTTCAATTGGCATTCTGGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD4B PubMed Links

Gene Details

Gene
MFSD4B
Gene Name
major facilitator superfamily domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153369.2 707 Silent Mutation TTC,TTT F95F NP_699200.2
XM_017011483.1 707 Intron XP_016866972.1

View Full Product Details