Product Details

SNP ID
rs139066335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138773701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGAGGGCGGAGCCAAGAGCGAC[A/C]CTGGGGGAGCAGGAAAAGGCGGGGC
Phenotype
MIM: 615353
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC28A PubMed Links

Gene Details

Gene
CCDC28A
Gene Name
coiled-coil domain containing 28A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015439.2 182 Silent Mutation ACA,ACC T9T NP_056254.1
XM_011535728.2 182 Silent Mutation ACA,ACC T9T XP_011534030.1
Gene
GVQW2
Gene Name
GVQW motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242740.1 182 Intron NP_001229669.1

View Full Product Details