Product Details

SNP ID
rs79592231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31138459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTCACTCTCAAAGAGCTCTC[A/G]GTAGGTTTGTAAATACTTAACTGAT
Phenotype
MIM: 605310 MIM: 613525 MIM: 613526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCHCR1 PubMed Links

Gene Details

Gene
CCHCR1
Gene Name
coiled-coil alpha-helical rod protein 1
There are no transcripts associated with this gene.

Gene
PSORS1C1
Gene Name
psoriasis susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014068.2 332 Missense Mutation AGC,GGC S15G NP_054787.2
Gene
PSORS1C2
Gene Name
psoriasis susceptibility 1 candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014069.2 332 Intron NP_054788.2

View Full Product Details