Product Details

SNP ID
rs139120707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132817345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTAAGGTCCCTGAGAATGGCTAT[A/G]ACAAATCTTAGTGATGGGAAACATT
Phenotype
MIM: 603660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS12 PubMed Links

Gene Details

Gene
RPS12
Gene Name
ribosomal protein S12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001016.3 Intron NP_001007.2
Gene
SNORA33
Gene Name
small nucleolar RNA, H/ACA box 33
There are no transcripts associated with this gene.

Gene
SNORD100
Gene Name
small nucleolar RNA, C/D box 100
There are no transcripts associated with this gene.

Gene
SNORD101
Gene Name
small nucleolar RNA, C/D box 101
There are no transcripts associated with this gene.

View Full Product Details