Product Details

SNP ID
rs139273641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158640712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCACAGCACCACCACTAGTGG[C/T]ACTTACCACACAGCAAATGCAGGCT
Phenotype
MIM: 601554 MIM: 613209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNLT1 PubMed Links

Gene Details

Gene
DYNLT1
Gene Name
dynein light chain Tctex-type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291602.1 Intron NP_001278531.1
NM_001291603.1 Intron NP_001278532.1
NM_006519.3 Intron NP_006510.1
Gene
SYTL3
Gene Name
synaptotagmin like 3
There are no transcripts associated with this gene.

Gene
TMEM181
Gene Name
transmembrane protein 181
There are no transcripts associated with this gene.

View Full Product Details