Product Details
- SNP ID
-
rs139309886
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:130133458 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGTTTTCATTGACCAGGTGCCC[A/G]GGAAGAACCCACCGTCCAGCAGGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
L3MBTL3
PubMed Links
Gene Details
- Gene
- L3MBTL3
- Gene Name
- l(3)mbt-like 3 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007102.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q633R |
NP_001007103.1 |
NM_032438.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
NP_115814.1 |
XM_005267161.4 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_005267218.1 |
XM_006715576.3 |
2132 |
Missense Mutation |
CAG,CGG |
Q693R |
XP_006715639.1 |
XM_006715578.3 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_006715641.1 |
XM_011536179.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_011534481.1 |
XM_011536180.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_011534482.1 |
XM_011536181.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_011534483.1 |
XM_011536183.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_011534485.1 |
XM_011536184.2 |
2132 |
Missense Mutation |
CAG,CGG |
Q658R |
XP_011534486.1 |
XM_017011355.1 |
2132 |
Missense Mutation |
CAG,CGG |
Q633R |
XP_016866844.1 |
XM_017011356.1 |
2132 |
Missense Mutation |
CAG,CGG |
Q633R |
XP_016866845.1 |
- Gene
- SAMD3
- Gene Name
- sterile alpha motif domain containing 3
View Full Product Details