Product Details

SNP ID
rs139328462
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27957322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGACCTTGGCTGGAGTTTCCACTC[C/T]TTGTGGTCTTCTTGATTTCTTACAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR2B6 PubMed Links

Gene Details

Gene
OR2B6
Gene Name
olfactory receptor family 2 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012367.1 82 Missense Mutation CTT,TTT L28F NP_036499.1

View Full Product Details