Product Details
- SNP ID
-
rs139415037
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149578868 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGGAGGAAAAAGAATATTTTGG[A/G]ATTGTCAGTGTAAGGATTTTAGTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GINM1
PubMed Links
Gene Details
- Gene
- GINM1
- Gene Name
- glycoprotein integral membrane 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138785.3 |
347 |
Silent Mutation |
GGA,GGG |
G108G |
NP_620140.1 |
View Full Product Details